missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH9A1 Recombinant Rabbit Monoclonal Antibody (6A7Q9)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | ALDH9A1 |
|---|---|
| Clone | 6A7Q9 |
| Concentration | 3.04 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284000
|
Invitrogen™
MA557911 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283713
![]() |
Invitrogen™
MA557912 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: GSVPTGMKIM EMSAKGIKPV TLELGGKSPL IIFSDCDMNN AVKGALMANF LTQGQVCCNG TRVFVQKEIL DKFTEEVVKQ TQRIKIGDPL LEDTRMGPLI NRPHLERVLG FVKVAKEQGA KVLCGGDIYV PEDPKLKDGY YMRPCVLTNC RDDMTCVKEE IFGPVMSILS FDTEAEVLER ANDTTFGLAA GVFTR.
ALDH9A1 belongs to the aldehyde dehydrogenase family of proteins. The protein has a high activity for oxidation of gamma-aminobutyraldehyde and other amino aldehydes. The enzyme catalyzes the dehydrogenation of gamma-aminobutyraldehyde to gamma-aminobutyric acid (GABA). This isozyme is a tetramer of identical 54-kD subunits.Specifications
| ALDH9A1 | |
| 3.04 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P49189, Q9JLJ2, Q9JLJ3 | |
| 223, 56752, 64040 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 256-450 of human ALDH9A1 (NP_000687.3). | |
| Antibody |
| 6A7Q9 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ALDH9A1 | |
| 4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; AA139417; Abaldh; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family member A1; aldehyde dehydrogenase 9 family, member A1; aldehyde dehydrogenase 9, subfamily A1; aldehyde dehydrogenase 9A1; aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; aldehyde dehydrogenase family 9, subfamily A1; ALDH4; ALDH7; ALDH9; Aldh9a1; E3; ESTM40; gamma-aminobutyraldehyde dehydrogenase; gamma-trimethylaminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABADH; TMABA-DH | |
| ALDH9A1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title