missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALDH9A1 Recombinant Rabbit Monoclonal Antibody (6A7Q9)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557911

 View more versions of this product

Product Code. 30284000

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GSVPTGMKIM EMSAKGIKPV TLELGGKSPL IIFSDCDMNN AVKGALMANF LTQGQVCCNG TRVFVQKEIL DKFTEEVVKQ TQRIKIGDPL LEDTRMGPLI NRPHLERVLG FVKVAKEQGA KVLCGGDIYV PEDPKLKDGY YMRPCVLTNC RDDMTCVKEE IFGPVMSILS FDTEAEVLER ANDTTFGLAA GVFTR.

ALDH9A1 belongs to the aldehyde dehydrogenase family of proteins. The protein has a high activity for oxidation of gamma-aminobutyraldehyde and other amino aldehydes. The enzyme catalyzes the dehydrogenation of gamma-aminobutyraldehyde to gamma-aminobutyric acid (GABA). This isozyme is a tetramer of identical 54-kD subunits.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALDH9A1
Recombinant Monoclonal
3.04 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P49189, Q9JLJ2, Q9JLJ3
ALDH9A1
Recombinant fusion protein containing a sequence corresponding to amino acids 256-450 of human ALDH9A1 (NP_000687.3).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
6A7Q9
Unconjugated
ALDH9A1
4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; AA139417; Abaldh; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family member A1; aldehyde dehydrogenase 9 family, member A1; aldehyde dehydrogenase 9, subfamily A1; aldehyde dehydrogenase 9A1; aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; aldehyde dehydrogenase family 9, subfamily A1; ALDH4; ALDH7; ALDH9; Aldh9a1; E3; ESTM40; gamma-aminobutyraldehyde dehydrogenase; gamma-trimethylaminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABADH; TMABA-DH
Rabbit
Protein A
RUO
223, 56752, 64040
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.