missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH2 Recombinant Rabbit Monoclonal Antibody (23GB4810), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ALDH2 |
|---|---|
| Clone | 23GB4810 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088418
![]() |
Invitrogen™
MA552750 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089096
|
Invitrogen™
MA552751 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALDH2 Recombinant Monoclonal antibody specifically detects ALDH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ALDH2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P05091, P11884, P47738 | |
| 11669, 217, 29539 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4810 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| acetaldehyde dehydrogenase 2; Ahd1; Ahd-1; Ahd5; Ahd-5; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH1; ALDH2; ALDH-E2; ALDHI; ALDM; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
| A synthesized peptide derived from human ALDH2 (250-315AA). | |
| Primary | |
| -20°C | |
| Aldh2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title