missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH2 Recombinant Rabbit Monoclonal Antibody (23GB4810), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552751
This item is not returnable.
View return policy
Description
ALDH2 belongs to the aldehyde dehydrogenase family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Asians have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Asians than among Caucasians could be related to the absence of the mitochondrial isozyme.
Specifications
| ALDH2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Aldh2 | |
| acetaldehyde dehydrogenase 2; Ahd1; Ahd-1; Ahd5; Ahd-5; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH1; ALDH2; ALDH-E2; ALDHI; ALDM; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; nucleus-encoded mitochondrial aldehyde dehydrogenase 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11669, 217, 29539 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4810 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05091, P11884, P47738 | |
| Aldh2 | |
| A synthesized peptide derived from human ALDH2 (250-315AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction