missing translation for 'onlineSavingsMsg'
Learn More

ALDH1L1 Antibody (2E7), Novus Biologicals™

Mouse Monoclonal Antibody

£146.00 - £385.00

Specifications

Antigen ALDH1L1
Clone 2E7
Concentration 1 mg/mL
Dilution Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000
Applications Western Blot, Immunohistochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18621548
View Documents
Novus Biologicals
NBP2-50033SS
0.025 mL
£146.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18630327
View Documents
Novus Biologicals
NBP2-50033
0.1 mL
£385.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

ALDH1L1
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Mouse
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction
50% PBS, 50% glycerol
10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF
Amino acids 402-902 of human ALDH1L1. [UniProt# O75891]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
2E7
Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000
Monoclonal
Purified
RUO
Human, Mouse, Rat
O75891
10840
IgG1
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.