missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L1 Antibody (2E7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50033
This item is not returnable.
View return policy
Description
ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| ALDH1L1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| O75891 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2E7 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| 10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
| Amino acids 402-902 of human ALDH1L1. [UniProt# O75891] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction | |
| 10840 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction