missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alcohol Dehydrogenase 1A Recombinant Rabbit Monoclonal Antibody (23GB4295), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Alcohol Dehydrogenase 1A |
|---|---|
| Clone | 23GB4295 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089942
![]() |
Invitrogen™
MA552678 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087682
|
Invitrogen™
MA552679 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alcohol Dehydrogenase 1A Recombinant Monoclonal antibody specifically detects Alcohol Dehydrogenase 1A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Alcohol Dehydrogenase 1A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P00329, P06757, P07327 | |
| 11522, 124, 24172 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4295 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ADC1; Adh; ADH, alpha subunit; ADH1; Adh-1; ADH1A; Adh1c; Adh-1e; Adh1-e; Adh-1t; Adh-1-t; Adh1-t; Adh1tl; Adh-3e; Adh3-e; ADH-A2; ADH-AA; AI194826; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I); alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 3, electrophoretic; alcohol dehydrogenase A subunit; alcohol dehydrogenase ADH1; Alcohol dehydrogenase I; Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase; O0947; YADH-1; YOL086C | |
| A synthesized peptide derived from human Alcohol Dehydrogenase (1-50AA). | |
| Primary | |
| -20°C | |
| ADH1A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts