missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alcohol Dehydrogenase 1A Recombinant Rabbit Monoclonal Antibody (23GB4295), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552678
This item is not returnable.
View return policy
Description
This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.
Specifications
| Alcohol Dehydrogenase 1A | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00329, P06757, P07327 | |
| Adh1, ADH1A | |
| A synthesized peptide derived from human Alcohol Dehydrogenase (1-50AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4295 | |
| Unconjugated | |
| ADH1A | |
| ADC1; Adh; ADH, alpha subunit; ADH1; Adh-1; ADH1A; Adh1c; Adh-1e; Adh1-e; Adh-1t; Adh-1-t; Adh1-t; Adh1tl; Adh-3e; Adh3-e; ADH-A2; ADH-AA; AI194826; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I); alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 3, electrophoretic; alcohol dehydrogenase A subunit; alcohol dehydrogenase ADH1; Alcohol dehydrogenase I; Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase; O0947; YADH-1; YOL086C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11522, 124, 24172 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction