missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2, p Ser474 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£203.00 - £324.00
Specifications
| Antigen | AKT2 (p Ser474) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207814
|
Novus Biologicals
NB100-81780-0.025ML |
0.025 mL |
£203.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18148779
|
Novus Biologicals
NB100-81780 |
0.05 mL |
£324.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT2 Polyclonal specifically detects AKT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AKT2 (p Ser474) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, mTOR Pathway, Phospho Specific | |
| P31751 | |
| 208 | |
| The antiserum was produced against synthesized phosphopeptide derived from human Akt2 around the phosphorylation site of serine 474 (Q-F-SP-Y-S). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Rat, Mouse, Human | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| AKT2 | |
| IgG | |
| Affinity Purified | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title