missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2, p Ser474 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81780-0.025ML
This item is not returnable.
View return policy
Description
AKT2 Polyclonal specifically detects AKT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AKT2 (p Ser474) | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| Rabbit | |
| 60 kDa | |
| 0.025 mL | |
| Cancer, mTOR Pathway, Phospho Specific | |
| 208 | |
| Rat, Mouse, Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P31751 | |
| AKT2 | |
| The antiserum was produced against synthesized phosphopeptide derived from human Akt2 around the phosphorylation site of serine 474 (Q-F-SP-Y-S). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Akt2 (specific to Phospho-Ser474) detects endogenous levels of Akt2 only when phosphorylated at serine 474. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction