missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1 Antibody (3A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£196.00
Specifications
| Antigen | AKT1 |
|---|---|
| Clone | 3A3 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AKT1 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 207 | |
| This AKT1 antibody is made to a purified recombinant fragment of human AKT1 expressed in E. coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 3A3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Signal Transduction | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| AKT1 | |
| IgG1 | |
| Unpurified | |
| 55.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title