missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1 Antibody (3A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-51602-0.025ml
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AKT1 | |
| Monoclonal | |
| Unconjugated | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| Mouse | |
| 55.7 kDa | |
| 0.025 mL | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Signal Transduction | |
| 207 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 3A3 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AKT1 | |
| This AKT1 antibody is made to a purified recombinant fragment of human AKT1 expressed in E. coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction