missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | AKR7A3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
AKR7A3 Polyclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot.Specifications
| AKR7A3 | |
| Polyclonal | |
| Rabbit | |
| O95154 | |
| 22977 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
| AKR7A3 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title