missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56791
This item is not returnable.
View return policy
Description
AKD1 Polyclonal specifically detects AKD1 in Human samples. It is validated for Western Blot.
Specifications
| AKD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| adenylate kinase domain containing 1, adenylate kinase domain containing 2, adenylate kinase domain-containing protein 1, Adenylate kinase domain-containing protein 2, AKD2, C6orf199, C6orf224, chromosome 6 open reading frame 199, chromosome 6 open reading frame 224, dJ70A9.1, FLJ16163, FLJ25791, FLJ34784, FLJ42177, MGC126763, MGC138153, MGC177059, MGC180194, MGC184281, MGC26954, RP1-70A9.1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Western clawed frog: 100%; Crab-eating macaque: 100%; Human: 100%; Chicken: 83%; Canine: 81%; Klebsiella sp. 1_1_55: 75%;. | |
| Human, Mouse, Rat, Pig, Canine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q5TCS8 | |
| AK9 | |
| Synthetic peptides corresponding to C6ORF199 The peptide sequence was selected from the N terminal of C6ORF199. Peptide sequence TSQEKTEEYPFADIFDEDETERNFLLSKPVCFVVFGKPGVGKTTLARYIT. | |
| 100 μL | |
| Protein Kinase | |
| 221264 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction