missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP3 Mouse, anti-Human, CoraLite Plus 488, Clone: 3C11H9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | AKAP3 |
|---|---|
| Clone | 3C11H9 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18036395
|
Proteintech
CL488-66255-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18097405
|
Proteintech
CL488-66255-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AKAP3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AKAP3. This antibody recognizes human, mouse antigen. The AKAP3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| AKAP3 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 10566 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3C11H9 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse | |
| A kinase anchor protein 3, AKAP 110, AKAP 3, AKAP110, AKAP3, Cancer/testis antigen 82, CT82, Fibrousheathin 1, Fibrousheathin I, FSP95, PRKA3, SOB1, Sperm oocyte binding protein | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title