Learn More
AKAP3 Mouse, anti-Human, CoraLite Plus 488, Clone: 3C11H9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66255-20UL
Description
The AKAP3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AKAP3. This antibody recognizes human, mouse antigen. The AKAP3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| AKAP3 | |
| 3C11H9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| A kinase anchor protein 3, AKAP 110, AKAP 3, AKAP110, AKAP3, Cancer/testis antigen 82, CT82, Fibrousheathin 1, Fibrousheathin I, FSP95, PRKA3, SOB1, Sperm oocyte binding protein | |
| 20 μL | |
| 10566 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |