missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress AkaLumine hydrochloride

Product Code. 16460957
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460957 50 mg 50mg
16454668 10 mg 10mg
16445506 100 mg 100mg
16407387 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16460957 Supplier MedChemExpress Supplier No. HY112641A
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AkaLumine hydrochloride is a luciferin analogue, with a Km of 2.06 μM for recombinant Fluc protein.

Specifications

Content And Storage -20°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen)
Quantity 50 mg
Shipping Condition Room temperature
Solubility H2O : 50 mg/mL (147.56 mM; Need ultrasonic) ∣ DMSO : 26.67 mg/mL (78.71 mM; Need ultrasonic)
Molecular Weight (g/mol) 338.85 g/mol
CAS 2558205-28-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.