missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF Antibody (3C11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £324.00
Specifications
| Antigen | AIF |
|---|---|
| Clone | 3C11 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18168317
|
Novus Biologicals
NBP2-50606 |
0.1 mL |
£324.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690927
|
Novus Biologicals
NBP2-50606-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AIF Monoclonal specifically detects AIF in Human samples. It is validated for Western Blot.Specifications
| AIF | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| AIFM1 | |
| IgG1 | |
| Protein A purified |
| 3C11 | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 9131 | |
| AIF-His fusion protein | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title