missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF Antibody (3C11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50606-0.025ml
This item is not returnable.
View return policy
Description
AIF Monoclonal specifically detects AIF in Human samples. It is validated for Western Blot.
Specifications
| AIF | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9131 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3C11 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AIFM1 | |
| AIF-His fusion protein | |
| 0.025 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction