missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHR Antibody (RPT9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB300-515-0.025ml
This item is not returnable.
View return policy
Description
AHR Monoclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Gel Super Shift Assays.
Specifications
| AHR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:100 | |
| P30561 | |
| AHR | |
| Synthetic peptide corresponding to residues R(12) K R R K P(17) V(22) K P I P A E G I K(31) of mouse AhR. | |
| 0.025 mL | |
| Apoptosis, Cancer, Hypoxia, Innate Immunity, Transcription Factors and Regulators | |
| 196 | |
| Human | |
| Purified |
| Western Blot | |
| RPT9 | |
| Unconjugated | |
| No buffer with 0.05% Sodium Azide | |
| Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Detects the Aryl Hydrocarbon Receptor (AHR). | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction