missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHR Antibody (RPT9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£174.00
Specifications
| Antigen | AHR |
|---|---|
| Clone | RPT9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:100 |
| Applications | Western Blot |
Description
AHR Monoclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Gel Super Shift Assays.Specifications
| AHR | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Hypoxia, Innate Immunity, Transcription Factors and Regulators | |
| No buffer with 0.05% Sodium Azide | |
| Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
| AHR | |
| IgG3 κ | |
| Protein A purified | |
| Detects the Aryl Hydrocarbon Receptor (AHR). |
| RPT9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P30561 | |
| 196 | |
| Synthetic peptide corresponding to residues R(12) K R R K P(17) V(22) K P I P A E G I K(31) of mouse AhR. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title