missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTR-2 Antibody (6C7), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | AGTR-2 |
|---|---|
| Clone | 6C7 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043353
|
Novus Biologicals
NBP3-26517-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043632
|
Novus Biologicals
NBP3-26517-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGTR-2 Monoclonal antibody specifically detects AGTR-2 in Human, Mouse samples. It is validated for Western Blot, ELISASpecifications
| AGTR-2 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor | |
| A synthesized peptide derived from Human AGTR-2 [UniProt P50052] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6C7 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, GPCR | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 186 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title