missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTR-2 Antibody (6C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26517-100ul
This item is not returnable.
View return policy
Description
AGTR-2 Monoclonal antibody specifically detects AGTR-2 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| AGTR-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 6C7 | |
| Western Blot 1:500-1:5000, ELISA | |
| Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor | |
| A synthesized peptide derived from Human AGTR-2 [UniProt P50052] | |
| 100 μL | |
| Apoptosis, GPCR | |
| 186 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction