missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGR3 Monoclonal Antibody (K1E028_18C10), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | AGR3 |
|---|---|
| Clone | K1E028_18C10 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30212060
|
Invitrogen™
MA555961 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30208481
|
Invitrogen™
MA556341 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: IAIKKEKRPP QTLSRGWGDD ITWVQTYEEG LFYAQKSKKP LMVIHHLEDC QYSQALKKVF AQNEEIQEMA QNKFIMLNLM HETTDKNLSP DGQYVPRIMF VDPSLTVRAD IAGRYSNRLY TYEPRDLPLL IENMKKALRL IQSEL
AGR3 Monoclonal antibody specifically detects AGR3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| AGR3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AG3; AG-3; AGR3; anterior gradient 3; anterior gradient 3 homolog; anterior gradient 3, protein disulphide isomerase family member; anterior gradient homolog 3; anterior gradient protein 3; anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; E030025L21Rik; Gm888; H PDIA18; HAG3; hAG-3; PDIA18; Protein disulfide isomerase family A, member 18; RGD1565406; UNQ642/PRO1272 | |
| Recombinant human AGR3 (immunogen range is 22-166). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| AGR3 |
| K1E028_18C10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8TD06 | |
| 155465 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title