missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGR3 Monoclonal Antibody (K1E028_18C10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556341
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: IAIKKEKRPP QTLSRGWGDD ITWVQTYEEG LFYAQKSKKP LMVIHHLEDC QYSQALKKVF AQNEEIQEMA QNKFIMLNLM HETTDKNLSP DGQYVPRIMF VDPSLTVRAD IAGRYSNRLY TYEPRDLPLL IENMKKALRL IQSEL
AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG-3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumour supressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumours. Its Xenopus homolog is associated with anteroposterior fate determination during early development.
Specifications
| AGR3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8TD06 | |
| AGR3 | |
| Recombinant human AGR3 (immunogen range is 22-166). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K1E028_18C10 | |
| Unconjugated | |
| AGR3 | |
| AG3; AG-3; AGR3; anterior gradient 3; anterior gradient 3 homolog; anterior gradient 3, protein disulphide isomerase family member; anterior gradient homolog 3; anterior gradient protein 3; anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; E030025L21Rik; Gm888; H PDIA18; HAG3; hAG-3; PDIA18; Protein disulfide isomerase family A, member 18; RGD1565406; UNQ642/PRO1272 | |
| Mouse | |
| Protein A | |
| RUO | |
| 155465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction