missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AGPAT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | AGPAT2 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067149
|
Novus Biologicals™
NBP3-30862-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067190
|
Novus Biologicals™
NBP3-30862-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGPAT2 Polyclonal antibody specifically detects AGPAT2 in Human samples. It is validated for ELISA,Western BlotSpecifications
| AGPAT2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| Phosphate buffered saline | |
| 10555 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot : 5-10 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 1-acylglycerol-3-phosphate O-acyltransferase 2, 1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acidacyltransferase, beta), 1-AGPAT 2,1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acidacyltransferase-beta), 1-AGPAT2,1-AGP acyltransferase 2,1-acyl-sn-glycerol-3-phosphate acyltransferase beta, Berardinelli-Seip congenital lipodystrophy, BSCL, BSCL1, EC 2.3.1.51, LPAAB, LPAAT-beta, Lysophosphatidic acid acyltransferase beta, lysophosphatidic acid acyltransferase-beta | |
| Synthetic peptide derived from the AGPAT2 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title