missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AGPAT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30862-100ug
This item is not returnable.
View return policy
Description
AGPAT2 Polyclonal antibody specifically detects AGPAT2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| AGPAT2 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, ELISA | |
| 1-acylglycerol-3-phosphate O-acyltransferase 2, 1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acidacyltransferase, beta), 1-AGPAT 2,1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acidacyltransferase-beta), 1-AGPAT2,1-AGP acyltransferase 2,1-acyl-sn-glycerol-3-phosphate acyltransferase beta, Berardinelli-Seip congenital lipodystrophy, BSCL, BSCL1, EC 2.3.1.51, LPAAB, LPAAT-beta, Lysophosphatidic acid acyltransferase beta, lysophosphatidic acid acyltransferase-beta | |
| Synthetic peptide derived from the AGPAT2 | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| 10555 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction