Learn More
AGMAT Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21899-1-AP-150UL
Description
The AGMAT antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human AGMAT. This antibody recognizes human, mouse, rat antigen. The AGMAT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| AGMAT | |
| 900 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| AGMAT, Agmatinase, Agmatinase mitochondrial, Agmatine ureohydrolase, AUH | |
| 150 μL | |
| 79814 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |