missing translation for 'onlineSavingsMsg'
Learn More

AGMAT Rabbit, anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen AGMAT
Concentration 900 μg/mL
Applications Western Blot, ELISA
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18079465
View Documents
Proteintech
21899-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18096885
View Documents
Proteintech
21899-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The AGMAT antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human AGMAT. This antibody recognizes human, mouse, rat antigen. The AGMAT antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

AGMAT
Western Blot, ELISA
Rabbit
79814
IgG
Store at -20°C. Stable for one year after shipment.
900 μg/mL
Liquid
Human, Mouse, Rat
AGMAT, Agmatinase, Agmatinase mitochondrial, Agmatine ureohydrolase, AUH
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.