missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The AGMAT antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human AGMAT. This antibody recognizes human, mouse, rat antigen. The AGMAT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | AGMAT |
| Applications | Western Blot, ELISA |
| Concentration | 900 μg/mL |
| Gene Symbols | AGMAT, Agmatinase, Agmatinase mitochondrial, Agmatine ureohydrolase, AUH |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 79814 |
| Target Species | Human, Mouse, Rat |
| Content And Storage | Store at -20°C. Stable for one year after shipment. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?