Learn More
RETSCH Agate Grinding Jars
Product Code. 10228230 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
For MM 200 Mixer Mill
For MM 301 Mixer mill
Capacity (Metric):
5 mL
10 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Capacity (Metric) unitSize
10228230 For MM 200 Mixer Mill 5 mL Each
10771581 For MM 200 Mixer Mill 10 mL Each
10457982 For MM 301 Mixer mill 10 mL Each
10427602 For MM 301 Mixer mill 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10228230 Supplier RETSCH Supplier No. 01.462.0113
Only null left
Add to basket
Add to basket
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Agate Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Product Type Grinding Jar
Capacity (Metric) 5 mL
For Use With (Equipment) For MM 200 Mixer Mill
Material Agate