missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Agate Comfort Grinding Jars

Use these grinding jars with Retsch™ ball mills. Retsch™ Agate Comfort Grinding Jars feature an O-type sealing for safe operation.

Brand:  RETSCH 01.462.0220

Code : 50

Additional Details : Weight : 3.50000kg

 View more versions of this product

Product Code. 10184822

  • £2257.60 / Each

Estimated Shipment: 31-03-2022
Add to basket

Description

Description

  • Available in a variety of sizes from 12 to 500mL
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications

Specifications

250mL
Agate
For PM 100, PM 400 and S 100
Comfort Grinding Jar
Videos
SDS
Documents
Certificates
View accessory products
Special Offers

Special Offers