missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | AGAP2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18117861
|
Novus Biologicals
NBP2-24554SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18746313
|
Novus Biologicals
NBP2-24554 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGAP2 Polyclonal specifically detects AGAP2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AGAP2 | |
| Western Blot 2.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 116986 | |
| A portion of amino acids 20-60 of human AGAP2 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Primate | |
| AGAP-2, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, centaurin-gamma-1, CENTG1, Cnt-g1, FLJ16430, GGAP2, GTP-binding and GTPase activating protein 2, GTP-binding and GTPase-activating protein 2, KIAA0167, Phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer, PIKEArf GAP with GTP-binding protein-like, ANK repeat and PH domains 2 | |
| AGAP2 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and monkey, 95% homologous in cow and 90% homologous in dog, rat and mouse. Additionally, it is 71% homologous in CRABP1, a 137 amino acid protein (CRABP2 is a 138 amino acid protein). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title