missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24554SS
This item is not returnable.
View return policy
Description
AGAP2 Polyclonal specifically detects AGAP2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AGAP2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| AGAP2 | |
| A portion of amino acids 20-60 of human AGAP2 was used as the immunogen. | |
| 0.025 mg | |
| Apoptosis | |
| 116986 | |
| Human, Primate | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 2.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| AGAP-2, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, centaurin-gamma-1, CENTG1, Cnt-g1, FLJ16430, GGAP2, GTP-binding and GTPase activating protein 2, GTP-binding and GTPase-activating protein 2, KIAA0167, Phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer, PIKEArf GAP with GTP-binding protein-like, ANK repeat and PH domains 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and monkey, 95% homologous in cow and 90% homologous in dog, rat and mouse. Additionally, it is 71% homologous in CRABP1, a 137 amino acid protein (CRABP2 is a 138 amino acid protein). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction