missing translation for 'onlineSavingsMsg'
Learn More

AG 825, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18096052
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18096052 50 mg 50mg
18023942 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18096052 Supplier Tocris Bioscience Supplier No. 1555/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective ErbB2 inhibitor

Selective ErbB2 inhibitor (IC50 values are 0.15 and 19 μM at ErbB2 and ErbB1 respectively). Preferentially triggers p38 MAP kinase-dependent apoptosis in androgen-independent prostate cancer cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149092-50-2
Molecular Formula C19H15N3O3S2
Molecular Weight (g/mol) 397.47
MDL Number MFCD01074971
InChI Key KXDONFLNGBQLTN-UHFFFAOYSA-N
Synonym tyrphostin ag 825, tyrphostin c15, 4-hydroxy-3-methoxy-5-benzothiazolylthiomethyl benzylidenecyanoacetamide, 5-benzothiazol-2-yl thiomethyl-4-hydroxy-3-methoxybenzylidenecyanoacetamide, 2e-3-3-1,3-benzothiazol-2-ylsulfanyl methyl-4-hydroxy-5-methoxyphenyl-2-cyanoprop-2-enamide, tocris-1555, biomolki_000008, biomolki2_000018, bmk1-b8, tyrphostin ag
PubChem CID 6091659
ChEBI CHEBI:75405
IUPAC Name 3-{3-[(1,3-benzothiazol-2-ylsulfanyl)methyl]-4-hydroxy-5-methoxyphenyl}-2-cyanoprop-2-enamide
SMILES COC1=CC(C=C(C#N)C(N)=O)=CC(CSC2=NC3=CC=CC=C3S2)=C1O

Specifications

Quantity 50 mg
Formula Weight 397.47
Percent Purity >98%
Chemical Name or Material AG 825
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.