missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-2/AGR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£183.00 - £386.00
Specifications
| Antigen | AG-2/AGR2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot reported in scientific literature (PMID 22072305), Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18232350
|
Novus Biologicals
NB110-17780SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18210171
|
Novus Biologicals
NB110-17780 |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| AG-2/AGR2 | |
| Western Blot reported in scientific literature (PMID 22072305), Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
| AGR2 | |
| IgG | |
| Affinity Purified | |
| Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse. |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| O95994 | |
| 10551 | |
| A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt# O95994] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts