missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-2/AGR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB110-17780SS
This item is not returnable.
View return policy
Description
AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AG-2/AGR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot reported in scientific literature (PMID 22072305), Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| O95994 | |
| AGR2 | |
| A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt# O95994] | |
| 0.025 mL | |
| Breast Cancer, Cancer | |
| 10551 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction