Learn More
Eppendorf™ Aerosol-Tight Rotor Lids
Product Code. 10386842 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
FA–45-24-11
FA–45-30-11
Rotor A-4-81
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10386842 FA–45-30-11 Each
10354902 Rotor A-4-81 Pack of 2
10305482 FA–45-24-11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10386842 Supplier Eppendorf™ Supplier No. 5490025806
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Providing secure closure for a variety of Eppendorf rotors.
  • Aluminim construction
  • Aerosol-tight
  • Autoclavable for 20 min at 121°C
TRUSTED_SUSTAINABILITY
Specifications
Type Aerosol-tight Lid
Material Aluminum
For Use With (Equipment) FA–45-30-11
Autoclavable Autoclavable (121°C for 20 min.)