missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aeromonas Medium

Product Code. 12789060
Change view
Click to view available options
Description:
Aeromonas Medium
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12789060 Aeromonas Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12789060 Supplier Thermo Scientific™ Supplier No. PO0325A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selectively isolate Aeromonas hydrophila from food, water and environmental specimens with this diagnostic medium

Selectively isolate Aeromonas hydrophila from food, water and environmental samples with the ready-to-use Thermo Scientific™ Oxoid™ Aeromonas Selective Agar. The medium is modification of XLD medium1 designed to support the growth of Aeromonas spp. and Plesiomonas spp. as well as usual Enterobacterioaceae. To improve the selectivity of the medium, ampicillin is added at recommended levels. The recommended level of ampicillin is well below the level which can cause inhibition of some strains of Aermonas.2

Aeromonas spp. occur widely in soil and water including untreated and chlorinated water, raw materials, raw foods and raw milk. Use Aeromonas Medium for selective isolation of Aeromonas hydrophila from environmental samples.

  • Recommended by the MAFF/DHS Steering Group on the Microbiological Safety of Food for detection and enumeration of Aeromonas hydrophila3.
  • Ready-to-use convenience of prepared media
The use of Aeromonas Agar for detection of Aeromonas spp. in tap water, bottled water and foods including meat, poultry, fish and seafoods has been reported by several workers. 4,5,6,7,8

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 10°C
Description Aeromonas Medium
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.