missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adropin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£170.00 - £386.00
Specifications
| Antigen | Adropin |
|---|---|
| Concentration | 1.11 mg/mL |
| Dilution | Western Blot 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18029775
|
Novus Biologicals
NBP1-26387SS |
0.025 mL |
£170.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18739543
|
Novus Biologicals
NBP1-26387 |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.Specifications
| Adropin | |
| Western Blot 2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 | |
| ENHO | |
| IgG | |
| Affinity Purified | |
| Human and mouse. 88% sequence identity with bovine protein. |
| 1.11 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q6UWT2 | |
| 375704 | |
| Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-Prot# Q6UWT2] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title