missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adropin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-26387
This item is not returnable.
View return policy
Description
Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Adropin | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 | |
| Rabbit | |
| 8 kDa | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism | |
| 375704 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 1.11 mg/mL | |
| Western Blot 2 ug/ml | |
| Q6UWT2 | |
| ENHO | |
| Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-Prot# Q6UWT2] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. 88% sequence identity with bovine protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction