missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRBK1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2F7A5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | ADRBK1 |
|---|---|
| Clone | 2F7A5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18007105
|
Proteintech
CL488-67067-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035215
|
Proteintech
CL488-67067-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ADRBK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADRBK1. This antibody recognizes human antigen. The ADRBK1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| ADRBK1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 156 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2F7A5 | |
| Western Blot | |
| Liquid | |
| Human | |
| ADRBK1, BARK, BARK1, Beta ARK 1, BETA ARK1, FLJ16718, GRK2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title