Learn More
ADRBK1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2F7A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67067-20UL
Description
The ADRBK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADRBK1. This antibody recognizes human antigen. The ADRBK1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| ADRBK1 | |
| 2F7A5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Western Blot | |
| 1000 μg/mL | |
| ADRBK1, BARK, BARK1, Beta ARK 1, BETA ARK1, FLJ16718, GRK2 | |
| 20 μL | |
| 156 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |