Learn More
GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells
GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells contain the human Adenosine A3 Receptor (ADORA3).
Brand: GeneBLAzer™ K1837
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: ADORA3
- Accession Number: NM_000677.2
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells contain the human Adenosine A3 Receptor (ADORA3) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line. The GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cells (Cat. no. K1537) contain a beta-lactamase reporter gene under control of the NFAT Response Element and a promiscuous G protein, Ga15. GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of NECA.
Specifications
Liquid Nitrogen | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
ADORA3-Gα15-NFAT-bla CHO-K1 cells, ADORA3-Gα15-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Dividing Cells |