missing translation for 'onlineSavingsMsg'
Learn More

ADO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ADO
Concentration 0.16 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16876114
View Documents
Proteintech
16479-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16866114
View Documents
Proteintech
16479-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human thiol dioxygenases include cysteine dioxygenase (CDO; MIM 603943) and cysteamine (2-amino ethane thiol) dioxygenase (ADO; EC 1.13.11.19). CDO adds 2 oxygen atoms to free cysteine, whereas ADO adds 2 oxygen atoms to free cysteamine to form hypotaurine.
Specifications

Specifications

ADO
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Human, Mouse
Q6PDY2, Q96SZ5
211488, 309732, 84890
ADO Fusion Protein Ag9598
Primary
-20°C
ADO
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
2 aminoethanethiol dioxygenase, ADO, C10orf22, Cysteamine dioxygenase
Ado
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.