Learn More
ADO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16479-1-AP-20UL
 View more versions of this product
Product Code. 16876114
  • £135.00 / 20µL
Description
Human thiol dioxygenases include cysteine dioxygenase (CDO; MIM 603943) and cysteamine (2-amino ethane thiol) dioxygenase (ADO; EC 1.13.11.19). CDO adds 2 oxygen atoms to free cysteine, whereas ADO adds 2 oxygen atoms to free cysteamine to form hypotaurine.
Specifications
Specifications
ADO
Polyclonal
Unconjugated
ADO
2 aminoethanethiol dioxygenase, ADO, C10orf22, Cysteamine dioxygenase
Rabbit
Antigen Affinity Chromatography
RUO
211488, 309732, 84890
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6PDY2, Q96SZ5
Ado
ADO Fusion Protein Ag9598
20 μL
Primary
Rat, Human, Mouse