Learn More
Adhesive Plate Sealer Film Polyester Non Toxic
Product Code. 12812447
Encompass
Product Code. 12812447 Supplier Brand Not Defined Supplier No. IBSA7740005
This item requires a price update prior to being ordered.
Request a Price for this item here.
A polyester film, with an acrylic based adhesive, gives you a non toxic plate cover which has excellent sealing properties, provides high clarity and UV resistance. The film offers a convenient and cost effective adhesive cover for long and short term storage. The tab allows quick removal and the film also creates a good reseal on top of a pierced foil seal. The backing is an 80 gauge draft paper, which allows easy peel removal, and the ink receptive top coating on the film is print friendly, notes and markings can be easily made on each plate sealer.