missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 6 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£163.00 - £254.00
Specifications
| Antigen | Adenylate Cyclase 6 |
|---|---|
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18538270
|
Novus Biologicals
NB100-75180-0.02mg |
0.02 mg |
£163.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18503503
|
Novus Biologicals
NB100-75180-0.025mg |
0.025 mg |
£254.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenylate Cyclase 6 Polyclonal specifically detects Adenylate Cyclase 6 in Human samples. It is validated for Western Blot.Specifications
| Adenylate Cyclase 6 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| O43306 | |
| 112 | |
| The immunogen for this antibody was a sequence from 1a.a. to 121a.a. from Adenylate cyclase type 6(a 1168a.a. long protein) | |
| Affinity Purified | |
| Adenylate Cyclase 6 |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Chicken | |
| Lipid and Metabolism, Vision | |
| PBS with No Preservative | |
| AC6, adenylate cyclase 6, adenylate cyclase type 6, Adenylate cyclase type VI, Adenylyl cyclase 6, ATP pyrophosphate-lyase 6, Ca(2+)-inhibitable adenylyl cyclase, DKFZp779F075, EC 4.6.1.1, KIAA0422 | |
| ADCY6 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title