missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 6 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75180-0.025mg
This item is not returnable.
View return policy
Description
Adenylate Cyclase 6 Polyclonal specifically detects Adenylate Cyclase 6 in Human samples. It is validated for Western Blot.
Specifications
| Adenylate Cyclase 6 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| O43306 | |
| ADCY6 | |
| The immunogen for this antibody was a sequence from 1a.a. to 121a.a. from Adenylate cyclase type 6(a 1168a.a. long protein) | |
| 0.025 mg | |
| Lipid and Metabolism, Vision | |
| 112 | |
| Human |
| Western Blot | |
| Unconjugated | |
| PBS with No Preservative | |
| AC6, adenylate cyclase 6, adenylate cyclase type 6, Adenylate cyclase type VI, Adenylyl cyclase 6, ATP pyrophosphate-lyase 6, Ca(2+)-inhibitable adenylyl cyclase, DKFZp779F075, EC 4.6.1.1, KIAA0422 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Adenylate Cyclase 6 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction