missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADCY6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118931
This item is not returnable.
View return policy
Description
ADCY6 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the adenylyl cyclase family of proteins, which are required for the synthesis of cyclic AMP. All members of this family have an intracellular N-terminus, a tandem repeat of six transmembrane domains separated by a cytoplasmic loop, and a C-terminal cytoplasmic domain. The two cytoplasmic regions bind ATP and form the catalytic core of the protein. Adenylyl cyclases are important effectors of transmembrane signaling pathways and are regulated by the activity of G protein coupled receptor signaling. This protein belongs to a small subclass of adenylyl cyclase proteins that are functionally related and are inhibited by protein kinase A, calcium ions and nitric oxide. A mutation in this gene is associated with arthrogryposis multiplex congenita.
Specifications
| ADCY6 | |
| Polyclonal | |
| Unconjugated | |
| ADCY6 | |
| AC6; ACVI; Adcy6; ADCYB; adenylate cyclase 6; adenylate cyclase type 6; Adenylate cyclase type VI; adenylyl cyclase 6; ATP pyrophosphate-lyase 6; ca(2+)-inhibitable adenylyl cyclase; DKFZp779F075; KIAA0422; LCCS8; mKIAA0422 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 112, 11512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O43306, Q01341 | |
| ADCY6 | |
| Recombinant protein Adenylate Cyclase 6. The antigen corresponds to amino acid range 314-501 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction